Tisk…
ISO safety signs - Wear laboratory coat, self adhesive
Dodavatel: Brady
311364 311365 311366 311367 311368 311415 311416 311417 311418 311419 311420 311421
BRDY311364EA 190 CZK
BRDY311364 BRDY311365 BRDY311366 BRDY311367 BRDY311368 BRDY311415 BRDY311416 BRDY311417 BRDY311418 BRDY311419 BRDY311420 BRDY311421
ISO safety signs - Wear laboratory coat, self adhesive
Značení bezpečnostní
Safety communication is vital in workplace accident prevention. Instructing and reminding employees and visitors that a laboratory coat must be worn is of crucial importance.
  • Increase safety using internationally recognised signs
  • Compliant with ISO 7010/ISO 3864
  • Designs are available in multiple top selling materials
  • Polyester over-laminated with a clear polyester
  • Permanent acrylic adhesive
Laboratory coats are a critical component of worker protection in both laboratory and clinical/patient care areas. In order to protect against accidental spill orMore Product Information