Tisk…
Label printers with peel function and vial label applicator, industrial thermal transfer benchtop, BradyPrinter i7100
Dodavatel: Brady
307652 307658 307653 307659
BRDY307652EA 191480 CZK
BRDY307652 116-0938 BRDY307653 BRDY307659
Label printers with peel function and vial label applicator, industrial thermal transfer benchtop, BradyPrinter i7100
Tiskárny Label Printers
The BradyPrinter i7100 industrial thermal transfer benchtop printer is ideal for customers requiring reliable high volume printing on lots of different materials within the aerospace, defence and mass transit, electronics, automotive, logistics, electrical, voice-datacom and LAB markets.
  • Prints labels and then applies them to wires or cables
  • Two step process
  • Laboratory labelling application
  • Lithium-manganese battery
  • Colour: Monocolour
It guarantees accurate printing on sleeves, tags, labels, tapes, cable markers and much more. This printer comes withMore Product Information
Objednat
Produktová specifikace
Environmentally Preferable
Připojení Ethernet, I/O connection, bluetooth (optional), WiFi (optional), serial, USB
Displej Colour touchscreen
Napájení AC adaptor
Metoda tisku Transfert thermique
Software Brady Workstation, Windows based driver for 3rd party software use
Podporovaný OS Windows® 7, Windows® 8, Windows® 10, Windows® Server 2012, Windows® Server 2012 R2, Windows® Server 2016
Rozsah teplot 5...40 °C