Tisk…
Safety lockout hasps, labelled
Dodavatel: Brady
265390 265392 265393
BRDY265390EA 4590 CZK
BRDY265390 BRDY265392 BRDY265393
Safety lockout hasps, labelled
Zámky bezpečnostní
A combination of lockout and tagout, these labelled hasps allow a single employee to lock out and service equipment without having separate hasps and tags.
  • Labels can be written on with permanent or non permanent marker pens
  • Made from tough 5052 anodised aluminium alloy
  • Six padlocks can be applied
Max. shackle diameter: 8,00 mm
Min. shackle clearance: 20,00 mm