Tisk…
Code Corporation CR2700 Barcode Scanner Accessories
Dodavatel: Brady
176699 176508
BRDY176699EA 3730 CZK
BRDY176699 BRDY176508
Code Corporation CR2700 Barcode Scanner Accessories
Systémy na čárové kódy
Accessories available for the Barcode Scanner CR2700.
Cable (BRDY176508) - Use this 6-foot straight cable for making USB to Micro USB connections. Reader termination: Micro USB. Host termination: USB Type A.
B27 lithium-ion battery (BRDY176699) - powers the CR2700 reader with a unique set of features that keep workflow interruptions to a minimum. Power gauge to track battery level. Quick-release latch allows for toolless swapping. The B27 battery can charge while installed in a CR2700 reader using the inductive charging station (CRA-A271).
Upozornení: Model BRDY176699 has lithium-More Product Information