Tisk…
Black Ink Cartridge for BradyJet™ J7300 Colour Label Printer, K True
Dodavatel: Brady
176505
BRDY176505EA 4750 CZK
BRDY176505
Black Ink Cartridge for BradyJet™ J7300 Colour Label Printer, K True
Inkoustové náplně
Get better results with BradyJet™ J7300 inks. Designed specifically for the J7300, BradyJet™ ink cartridges produce vibrant colours, last long, and deliver value in every drop.
  • K True Black ink cartridge delivers a deep black, darker tones, reliable barcode readability, and sharp text
  • Works in tandem with separate CMY ink cartridge to mix more accurate, durable colours
  • For use with the BradyJet™ J7300 colour label printer
Ink works with enhanced J7300 label material topcoat to dry instantly and won’t smudge, smear, or spot. Self-contained cartridgeMore Product Information