Tisk…
Repositionable Multi-Purpose Vinyl Labels for M710 and BMP71
Dodavatel: Brady
173288 173221 173258 173257 173289 173222
BRDY173288EA 2980 CZK
BRDY173288 BRDY173221 BRDY173258 BRDY173257 BRDY173289 BRDY173222
Repositionable Multi-Purpose Vinyl Labels for M710 and BMP71
Štítky
Repositionable vinyl film (B-581) is a durable, flexible, high gloss material ideal for indoor marking where frequent repositioning is required.
  • Special aggressive adhesive stays in place but removes cleanly without leaving adhesive residue
  • Ideal for applications like 5S and inventory
  • Vinyl film can be thermal transfer printed
  • Gloss finish
  • Average outdoor durability is five years
  • Service temperature of −40 to +82 °C (−40 to +180 °F)
Labels are ideal for facility identification, general identification, and safety identification applications.