Tisk…
Brady® BS 5609 Marine Labels for BradyJet™ J7300 Colour Label Printer
Dodavatel: Brady
176619 170469 176620 176586 170470
BRDY176619EA 10190 CZK
BRDY176619 BRDY170469 BRDY176620 BRDY176586 BRDY170470
Brady® BS 5609 Marine Labels for BradyJet™ J7300 Colour Label Printer
Štítky
PP labels B-2585 are certified for BS 5609 section 2 and 3 to ensure identification stays compliant, stuck, and seen during marine transport. For use on smooth surfaces.
  • Inkjet receptive polypropylene material
  • Aggressive adhesive
  • Designed for indoor/outdoor use
  • Abrasion-resistant, UV-resistant, water-resistant, weather-resistant
  • Matt finish
Ideal for GHS, hazardous materials, drum labelling, marine, transportation (DOT), storage, and pharmaceutical labelling. Built-in LabelSense™ technology enables cartridge and printer communication for fastMore Product Information