Tisk…
Floor corner signs, PE
Dodavatel: Brady
104430 104431 104432
BRDY104430EA 1170 CZK
BRDY104430 BRDY104431 BRDY104432
Floor corner signs, PE
Značení bezpečnostní
Corner marks are made from surface printed polyester with a rigid, clear polyester overlaminate.
  • Available in several colours