Tisk…
Labels for thermal transfer printers, type B-438
Dodavatel: Brady
030182
BRDY030182EA 8650 CZK
BRDY030182
Labels for thermal transfer printers, type B-438
Štítky
  • Fuels or oils resistant
  • High and low temperatures resistant
  • Solvents or chemicals resistant
  • Fraud resistant
Certifikace: UL recognised.