Tisk…
Fine dosage syringes, Injekt®-F Duo
Dodavatel: B. Braun
9166033V
BRAU9166033VEA 1450 CZK
BRAU9166033V
Fine dosage syringes, Injekt®-F Duo
Stříkačky Střílkačky zdravotnické
PP/PE fine dosage syringes, 1 ml, green plunger, two-piece, with single use hypodermic Sterican® needle. Smear-resistant, black graduation for ideal readability. Latex-, PVC- and silicone oil-free.
  • Highly transparent barrel
  • With displacement spike - no dead space, no medication wasted
  • Minimal residual volume
  • Secure plunger stopper for trouble-free filling to maximum volume
  • Centric Luer fitting
Balení: Dispenser box of 100 units.