Tisk…
Five-fold tap banks, Discofix® C
Dodavatel: B. Braun
16609C
BRAU16609CEA 17210 CZK
BRAU16609C
Five-fold tap banks, Discofix® C
Infuze
Five-fold tap banks are used for administration of medicines and medicines such as anesthetics, cytostatics, antibiotics, narcoleptics, immunosuppressants and lipids.
  • Leaks caused by stress cracks, contamination and air embolism are prevented
  • Easy access through the rotary adapter for injections and aspirations
  • Control of the 45° position by 'clicking'
  • Pressure resistant up to 2 bar
  • PVC and DEHP-free