Tisk…
Anti-EDRF1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Dodavatel: Bioss
BS-9737R-A647
BOSSBS-9737R-A647EA 25430 CZK
BOSSBS-9737R-A647
Anti-EDRF1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
The tetratricopeptide repeat (TPR) motif is a degenerate, 34 amino acid sequence found in many proteins and acts to mediate protein-protein interactions in various pathways. At the sequence level, there can be up to 16 tandem TPR repeats, each of which has a helix-turn-helix shape that stacks on other TPR repeats to achieve ligand binding specificity. EDRF1 (erythroid differentiation-related factor 1), also known as C10orf137 (chromosome 10 open reading frame 137), is a 1,238 amino acid protein containing two TPR repeats. Localizing to nucleus, EDRF1 is involved in transcriptional activationMore Product Information