Tisk…
Anti-FAM96B Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Dodavatel: Bioss
BS-9729R-A555
BOSSBS-9729R-A555EA 25430 CZK
BOSSBS-9729R-A555
Anti-FAM96B Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
Chromosome 16 encodes over 900 genes in approximately 90 million base pairs, makes up nearly 3% of human cellular DNA and is associated with a variety of genetic disorders. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth. The rare disorder Rubinstein-Taybi syndrome is also associated with chromosome 16, though through the CREBBP gene which encodes a critical CREB binding protein. Signs of Rubinstein-Taybi include mental retardation and predisposition to tumor growthMore Product Information
Objednat
Produktová specifikace
Symbol antigenu FAM96B
Environmentally Preferable
Název antigenu Family With Sequence Similarity 96, Member B
Synonyma antigenu CGI-128|Mitotic spindle-associated MMXD complex subunit MIP18|Protein FAM96B|MIP18|MSS19-interacting protein of 18 kDa|MIP18_HUMAN.
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 555
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 51647
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at –20 °C for 12 months
Transportní teplota 4°C