Tisk…
Anti-CHPT1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Dodavatel: Bioss
BS-9725R-A750
BOSSBS-9725R-A750EA 25430 CZK
BOSSBS-9725R-A750
Anti-CHPT1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
CHPT1, also known as AAPT1-like protein and Diacylglycerol cholinephosphotransferase 1, is a 406 amino acid multi-pass membrane protein that is localised to the golgi apparatus. By catalyzing the phosphatidylcholine biosynthesis from CDP-choline, it plays an essential role in the formation and maintenance of vesicular membranes. CHPT1 is most abundant in testis, as well as small intestine, heart, colon, spleen and prostate. Expression of CHPT1 is increased in cancerous breast cells as compared to normal breast cell lines and it has been determined that the CHPT1 gene exhibits mutationsMore Product Information