Tisk…
Anti-SYNE4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Dodavatel: Bioss
BS-9682R-FITC
BOSSBS-9682R-FITCEA 38440 CZK
BOSSBS-9682R-FITC
Anti-SYNE4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
C19orf46 is a 404 amino acid single-pass type IV membrane protein that belongs to the nesprin family. C19orf46 partcipates in the establishment of secretory epithelial morphology by inducing kinesin-dependent apical migration of the centrosome and Golgi apparatus as well as basal localization of the nucleus. C19orf46 interacts with UKHC and KCL1, and exists as two alternatively spliced isoforms. C19orf46 contains a KASH domain, which consists of a transmembrane motif, mediates nuclear enevelop targeting and binds to the SUN domain of SUN1 and SUN2. C19orf46 is encoded by a gene located onMore Product Information