Tisk…
Anti-C19ORF45 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Dodavatel: Bioss
BS-9681R-A647
BOSSBS-9681R-A647EA 25430 CZK
BOSSBS-9681R-A647
Anti-C19ORF45 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
C19orf45 is a 505 amino acid protein that is encoded by a gene located on human chromosome 19. Chromosome 19 consists of approximately 63 million bases and makes up over 2% of human genomic DNA. Chromosome 19 includes a diversity of interesting genes and is recognized for having the greatest gene density of the human chromosomes. It is the genetic home for a number of immunoglobulin superfamily members including the killer cell and leukocyte Ig-like receptors, a number of ICAMs, the CEACAM and PSG family, and Fcα receptors. Key genes for eye color and hair color also map to chromosome 19.More Product Information
Objednat
Produktová specifikace
Symbol antigenu C19ORF45
Název antigenu Chromosome 19 open reading frame 45
Environmentally Preferable
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 647
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 374877
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at –20 °C for 12 months
Transportní teplota 4°C