Tisk…

Anti-NPAP1 Rabbit Polyclonal Antibody (Cy5.5®)

Dodavatel: Bioss
BS-9625R-CY5.5
BOSSBS-9625R-CY5.5EA 25630 CZK
BOSSBS-9625R-CY5.5
Anti-NPAP1 Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
C15orf2 may be involved in spermatogenesis. Testis-specific in adults.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NPAP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR