Tisk…
Anti-FECH Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Dodavatel: Bioss
BS-9521R-FITC
BOSSBS-9521R-FITCEA 25630 CZK
BOSSBS-9521R-FITC
Anti-FECH Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Ferrochelatase catalyzes the insertion of the ferrous form of iron into protoporphyrin IX in the heme synthesis pathway, and is localised in the mitochondrion. Defects in ferrochelatase are associated with protoporphyria. Two transcript variants encoding different isoforms have been found for this gene. Porphyrias are a group of inherited or acquired disorders of certain enzymes in the heme biosynthetic pathway (also called porphyrin pathway). They are broadly classified as hepatic porphyrias or erythropoietic porphyrias, based on the site of the overproduction and mainly accumulation of theMore Product Information