Tisk…
Anti-SOSSC Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Dodavatel: Bioss
BS-9491R-A750
BOSSBS-9491R-A750EA 25430 CZK
BOSSBS-9491R-A750
Anti-SOSSC Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
The SOSS (Sensor of single-strand DNA) complex consists of multiple proteins that promote DNA repair and G2/M checkpoint downstream of the MRN (Mre11, Rad50 and Nbs1) complex. The complex is composed of SSBP1, INTS3 and C9orf80. Specifically, the SOSS complex binds to ssDNA at DNA lesions that influences diverse endpoints in the cellular DNA damage response. The complex is required for efficient homologous recombination-dependent repair of double-stranded breaks and ATM-dependent Signalling pathways. C9orf80, also known as SOSS complex subunit C and Single-stranded DNA-binding protein-More Product Information