Tisk…
Anti-TSPAN15 Rabbit Polyclonal Antibody (Cy7®)
Dodavatel: Bioss
BS-9425R-CY7
BOSSBS-9425R-CY7EA 23100 CZK
BOSSBS-9425R-CY7
Anti-TSPAN15 Rabbit Polyclonal Antibody (Cy7®)
Protilátky
NET-7, also known as TSPAN15 (tetraspanin 15) or TM4SF15 (transmembrane 4 superfamily member 15), is a 294 amino acid multi-pass membrane protein that belongs to the transmembrane 4 superfamily, also known as the tetraspanin family. Members of the tetraspanin family are cell-surface proteins that are characterized by the presence of four hydrophobic domains and mediate signal transduction events that play a role in the regulation of cell development, activation, growth, motility, differentiation, and cancer. Considered molecular facilitators, tetraspanin proteins may regulate vesicle fusionMore Product Information
Objednat
Produktová specifikace
Symbol antigenu TSPAN15
Environmentally Preferable
Název antigenu Tetraspanin 15
Synonyma antigenu Tetraspanin 15|Tetraspanin-15|TSPAN15.|TSN15_HUMAN|Transmembrane 4 superfamily member 15|Tetraspan NET-7|Tspan-15
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy7®
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 23555
Izotyp IgG
Imunochemie Yes
Průtoková cytometrie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at –20 °C for 12 months
Transportní teplota 4°C