Tisk…

Anti-MAP7D1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Dodavatel: Bioss
BS-9314R-A680
BOSSBS-9314R-A680EA 25430 CZK
BOSSBS-9314R-A680
Anti-MAP7D1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Protilátky
MAP7D1 (MAP7 Domain Containing 1) is a Protein-Coding gene. Gene Ontology (GO) annotations related to this gene include structural molecule activity. An important paralog of this gene is MAP7.

Type: Primary
Antigen: MAP7D1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/841
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR