Tisk…
Anti-ART3 Rabbit Polyclonal Antibody (Cy3®)
Dodavatel: Bioss
BS-9103R-CY3
BOSSBS-9103R-CY3EA 25630 CZK
BOSSBS-9103R-CY3
Anti-ART3 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
Mono-ADP-ribosylation is one of the posttranslational protein modifications regulating cellular metabolism (e.g. nitrogen fixation) in prokaryotes. Mono-ADP-ribosylation is a process in which the ADP-ribose moiety of nicotinamide adenine dinucleotide is transferred to an acceptor amino acid. Five mammalian ADP-ribosyltransferases (ART1-ART5) have been cloned, and expression is restricted to tissues such as cardiac and skeletal muscle, leukocytes, brain and testis. ART3 (ADP-ribosyltransferase 3), also known as Ecto-ADP-ribosyltransferase 3, is a testis specific membrane protein that does notMore Product Information