Tisk…
Anti-AGXT2L2 Rabbit Polyclonal Antibody (Cy5.5®)
Dodavatel: Bioss
BS-9095R-CY5.5
BOSSBS-9095R-CY5.5EA 25630 CZK
BOSSBS-9095R-CY5.5
Anti-AGXT2L2 Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
Members of the class-III pyridoxal-phosphate-dependent aminotransferase family, such as AGXT2, catalyze the conversion of glyoxylate to glycine using L-alanine as the amino donor. AGXT2 protects from asymmetric dimethylarginine (ADMA)-induced inhibition in nitric oxide (NO) production. Elevated blood concentrations of ADMA, a methyl derivate of the amino acid arginine and an endogenous inhibitor of nitric oxide (NO) synthase, is produced by the physiological degradation of methylated proteins and is found in association with diabetes, hypertension, congestive heart failure andMore Product Information