Tisk…
Anti-PDZD2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Dodavatel: Bioss
BS-9037R-A555
BOSSBS-9037R-A555EA 25430 CZK
BOSSBS-9037R-A555
Anti-PDZD2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
Proteins containing PDZ domains have been shown frequently to bind the C-termini of transmembrane receptors or ion channels. They have also been shown to bind to other PDZ domain proteins and could possibly be involved in intracellular signalling. PDZK3 contains six PDZ domains and shares sequence similarity with pro-interleukin-16 (pro-IL-16). Like pro-IL-16, the encoded protein localizes to the endoplasmic reticulum and is thought to be cleaved by a caspase to produce a secreted peptide containing two PDZ domains. In addition, the PDZK3 gene is upregulated in primary prostate tumors andMore Product Information