Tisk…

Anti-AKAP2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BS-9028R-A647
BOSSBS-9028R-A647EA 25430 CZK
BOSSBS-9028R-A647
Anti-AKAP2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
Binds to regulatory subunit (RII) of protein kinase A. May be involved in establishing polarity in signaling systems or in integrating PKA-RII isoforms with downstream effectors to capture, amplify and focus diffuse, trans-cellular signals carried by cAMP (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AKAP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR