Tisk…
Anti-CECR6 Rabbit Polyclonal Antibody (Cy5.5®)
Dodavatel: Bioss
BS-9006R-CY5.5
BOSSBS-9006R-CY5.5EA 25630 CZK
BOSSBS-9006R-CY5.5
Anti-CECR6 Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
Adenosine deaminase is an enzyme that is present in most tissues and exists predominantly as a monomer, although in some tissues it is associated with adenosine deaminase-binding protein. Adenosine deaminase degrades extracellular adenosine, which is toxic for lymphocytes. A novel family of growth factors that share sequence similarity to adenosine deaminase has been identified. The cat eye syndrome critical region protein (CECR) family includes CECR1, CECR2, CECR3, CECR4, CECR5, CECR6, CECR7, CECR8 and CECR9. The genes encoding CECR proteins are candidates for Cat Eye Syndrome (CES), aMore Product Information