Tisk…
Anti-BRMS1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Dodavatel: Bioss
BS-8686R-A647
BOSSBS-8686R-A647EA 25430 CZK
BOSSBS-8686R-A647
Anti-BRMS1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
Breast cancer metastasis-suppressor 1 (BRMS1) is 246 amino acid protein that acts as a mediator of metastasis suppression in several types of cancer including ovarian, lung, bladder, and murine mammary. BRMS1 mRNA is expressed in various tissues, including ovary, prostate, testis, and colon, but the protein is primarily detected in term placenta. BRMS1 suppresses metastasis without inhibiting tumorigenicity by modifying several metastasis-associated phenotypes. BRMS1 may participate in transcriptional regulation by binding to the mSin3/histone deacetylase complex. The expression of BRMS1 inMore Product Information
Objednat
Produktová specifikace
Symbol antigenu BRMS1
Environmentally Preferable
Název antigenu Breast Cancer Metastasis-suppressor 1
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 647
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at –20 °C for 12 months
Transportní teplota 4°C