Tisk…

Anti-GIPC-2 Rabbit Polyclonal Antibody (Cy5®)

Dodavatel: Bioss
BS-8586R-CY5
BOSSBS-8586R-CY5EA 25630 CZK
BOSSBS-8586R-CY5
Anti-GIPC-2 Rabbit Polyclonal Antibody (Cy5®)
Protilátky
GIPC2 is a 315 amino acid protein that localizes to the cytoplasm and contains one PDZ domain. Expressed at high levels in kidney and colon and at lower levels in adult liver, GIPC2 interacts with SEMA5A and is thought to function as a scaffold protein, possibly modulating cell adhesion and growth factor signaling and playing a role in tumorigenesis. The gene encoding GIPC2 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GIPC-2
Clonality:More Product Information
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR