Tisk…

Anti-TLR5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Bioss
BS-8539R-FITC
BOSSBS-8539R-FITCEA 25630 CZK
BOSSBS-8539R-FITC
Anti-TLR5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Luciferase from the firefly has become one of the more widely used reporter proteins for the study of gene expression. Luciferase catalyzes a bioluminescent reaction which requires the substrate luciferin as well as Mg2+ and ATP. Mixing these reagents with the cell extract containing luciferase, results in a flash of light that decays rapidly. This light can be detected by a luminometer. The total light emission is proportional to the luciferase activity of the sample.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TLR5
Clonality: Polyclonal
Clone:
Conjugation: FITC (More Product Information
Objednat

Produktová specifikace

Symbol antigenu TLR5
Název antigenu Toll-like Receptor 5
Environmentally Preferable
Synonyma antigenu TIL3|Toll-like receptor 5|Toll/interleukin-1 receptor-like protein 3
Druh protilátky Primární
Klonovalita Polyclonal
Spojení FITC (Fluorescein Isothiocyanate)
Reaktivita Multiple
Hostitel Rabbit
Identifikační číslo genu 7100
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at –20 °C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR