Tisk…

Anti-TLR5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Dodavatel: Bioss
BS-8539R-A488
BOSSBS-8539R-A488EA 25430 CZK
BOSSBS-8539R-A488
Anti-TLR5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
Luciferase from the firefly has become one of the more widely used reporter proteins for the study of gene expression. Luciferase catalyzes a bioluminescent reaction which requires the substrate luciferin as well as Mg2+ and ATP. Mixing these reagents with the cell extract containing luciferase, results in a flash of light that decays rapidly. This light can be detected by a luminometer. The total light emission is proportional to the luciferase activity of the sample.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TLR5
Clonality: Polyclonal
Clone:
Conjugation: AlexaMore Product Information
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR