Tisk…
Anti-UBE2D2 Rabbit Polyclonal Antibody (Cy5.5®)
Dodavatel: Bioss
BS-8348R-CY5.5
BOSSBS-8348R-CY5.5EA 25630 CZK
BOSSBS-8348R-CY5.5
Anti-UBE2D2 Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
Ubiquitin is an abundant, highly conserved protein found in all eukaryotic cells either free or covalently attached to cellular proteins. The primary function of ubiquitin in mammalian systems is to clear abnormal, foreign, and improperly folded proteins by targeting them for proteosome degradation. UBE2D proteins, including UBE2D1 (ubiquitin-conjugating enzyme E2D1 or UBC5A), UBE2D2 (ubiquitin-conjugating enzyme E2D2 or UBC5B) and UBE2D3 (ubiquitin-conjugating enzyme E2D3 or UBC5C), are E2 ubiquitin-conjugating enzymes that catalyze the ubiquitination of I˚Bå in a phosphorylation and SCFB-More Product Information