Tisk…
Anti-HDHD3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Dodavatel: Bioss
BS-8318R-FITC
BOSSBS-8318R-FITCEA 25630 CZK
BOSSBS-8318R-FITC
Anti-HDHD3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
HDHD3 (haloacid dehalogenase-like hydrolase domain containing 3) is a 251 amino acid protein that belongs to the HAD-like hydrolase superfamily, which contains a group of hydrolase enzymes that differ from the å/∫ hydrolase family based on structure. This family of hydrolase enzymes includes L-2-haloacid dehalogenase, epoxide hydrolases and phosphatases. HDHD3 is encoded by a gene located on human chromosome 9q32. Chromosome 9 consists of about 145 million bases, 4% of the human genome and encodes nearly 900 genes. Thought to play a role in gender determination, deletion of the distalMore Product Information
Objednat
Produktová specifikace
Symbol antigenu HDHD3
Název antigenu haloacid dehalogenase-like hydrolase domain containing 3
Environmentally Preferable
Synonyma antigenu 2810435D12Rik|C9orf158
Druh protilátky Primární
Klonovalita Polyclonal
Spojení FITC (Fluorescein Isothiocyanate)
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 81932
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at –20 °C for 12 months
Transportní teplota 4°C