Tisk…
Anti-DHRS7 Rabbit Polyclonal Antibody
Dodavatel: Bioss
BS-8262R
BOSSBS-8262REA 20650 CZK
BOSSBS-8262R
Anti-DHRS7 Rabbit Polyclonal Antibody
Protilátky
DHRS7 (dehydrogenase/reductase (SDR family) member 7), also known as SDR34C1, CGI-86 or retSDR4, is a 339 amino acid member of the SDR family. Like other members of the SDR family, DHRS7 contains a cofactor-binding Rossman-fold domain and is thought to catalyze the oxidation and reduction of a variety of substrates such as steroids and retinoids. DHRS7 exists as two alternatively spliced isoforms that are encoded by a gene located on human chromosome 14, which houses over 700 genes and comprises nearly 3.5% of the human genome. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which isMore Product Information