Tisk…
Anti-NSD1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Dodavatel: Bioss
BS-8170R-HRP
BOSSBS-8170R-HRPEA 25630 CZK
BOSSBS-8170R-HRP
Anti-NSD1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
This gene encodes a protein containing a SET domain, 2 LXXLL motifs, 3 nuclear translocation signals (NLSs), 4 plant homeodomain (PHD) finger regions, and a proline-rich region. The encoded protein enhances androgen receptor (AR) transactivation, and this enhancement can be increased further in the presence of other androgen receptor associated coregulators. This protein may act as a nucleus-localized, basic transcriptional factor and also as a bifunctional transcriptional regulator. Mutations of this gene have been associated with Sotos syndrome and Weaver syndrome. One version of childhoodMore Product Information