Tisk…

Anti-TBL1Y Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Dodavatel: Bioss
BS-8164R-A350
BOSSBS-8164R-A350EA 25430 CZK
BOSSBS-8164R-A350
Anti-TBL1Y Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
FunctionF-box-like protein involved in the recruitment of the ubiquitin/19S proteasome complex to nuclear receptor-regulated transcription units. Plays an essential role in transcription activation mediated by nuclear receptors. Probably acts as integral component of corepressor complexes that mediates the recruitment of the 19S proteasome complex, leading to the subsequent proteasomal degradation of transcription repressor complexes, thereby allowing cofactor exchange.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TBL1Y
Clonality: Polyclonal
Clone:
Conjugation: AlexaMore Product Information
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR