Tisk…

Anti-CA6 Rabbit Polyclonal Antibody (Cy5.5®)

Dodavatel: Bioss
BS-7983R-CY5.5
BOSSBS-7983R-CY5.5EA 25630 CZK
BOSSBS-7983R-CY5.5
Anti-CA6 Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
The protein encoded by this gene is one of several isozymes of carbonic anhydrase. This protein is found only in salivary glands and saliva and protein may play a role in the reversible hydratation of carbon dioxide though its function in saliva is unknown. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CA6
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR