Tisk…

Anti-ASNA1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Bioss
BS-7972R-FITC
BOSSBS-7972R-FITCEA 25630 CZK
BOSSBS-7972R-FITC
Anti-ASNA1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
ASNA1 is the human homolog of the E.coli arsA gene which is an ATPase, and is the catalytic component of a multisubunit oxyanion pump responsible for resistance to arsenicals and antimonials.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ASNA1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR