Tisk…

Anti-ABHD14B Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Dodavatel: Bioss
BS-7945R-A488
BOSSBS-7945R-A488EA 25430 CZK
BOSSBS-7945R-A488
Anti-ABHD14B Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
ABHD14B has hydrolase activity towards p-nitrophenyl butyrate (in vitro). It may activate transcription. There are 2 isoforms produced by alternative splicing.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABHD14B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR