Tisk…

Anti-PPM1G Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Bioss
BS-7930R-HRP
BOSSBS-7930R-HRPEA 25630 CZK
BOSSBS-7930R-HRP
Anti-PPM1G Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Tissue specificity:Widely expressed. Most abundant in testis, skeletal muscle, and heart.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: PPM1G
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR