Tisk…

Anti-PPM1G Rabbit Polyclonal Antibody (Cy5®)

Dodavatel: Bioss
BS-7930R-CY5
BOSSBS-7930R-CY5EA 25630 CZK
BOSSBS-7930R-CY5
Anti-PPM1G Rabbit Polyclonal Antibody (Cy5®)
Protilátky
Tissue specificity:Widely expressed. Most abundant in testis, skeletal muscle, and heart.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PPM1G
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR