Tisk…

Anti-Intra Acrosomal Protein Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Dodavatel: Bioss
BS-7866R-A750
BOSSBS-7866R-A750EA 25430 CZK
BOSSBS-7866R-A750
Anti-Intra Acrosomal Protein Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
This gene encodes a testis-specific, differentiation antigen, acrosomal vesicle protein 1, that arises within the acrosomal vesicle during spermatogenesis, and is associated with the acrosomal membranes and matrix of mature sperm. The acrosomal vesicle protein 1 may be involved in sperm-zona binding or penetration. Alternatively spliced transcript variants have been described.

Type: Primary
Antigen: Intra Acrosomal Protein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 171-265/265
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Produktová specifikace

Název antigenu Intra Acrosomal Protein
Environmentally Preferable
Synonyma antigenu Sp10|SP10|ASPX_HUMAN|SP 10|D11S4365|Sperm protein 10|SP-10|SPACA2|Acrosomal protein SP-10|Acrosomal protein SP 10|Acrosomal vesicle protein 1|Msa63|ACRV1
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 750
Reaktivita Mouse
Hostitel Rabbit
Identifikační číslo genu 6676
Izotyp IgG
Western blot Yes
Imunochemie Yes
ImunoFluorescence Yes
Modifikace Unmodified
Epitop 171-265/265
Imunogen KLH conjugated synthetic peptide derived from human Intra Acrosomal Protein
Formulář Liquid
Molekulární hmotnost 27 kDa
Čištění Purified by Protein A
Koncentrace 1 µg/µl
Skladovací pufr Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Skladovací teplota Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transportní teplota 4 °C
Testované aplikace ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR