Tisk…

Anti-Intra Acrosomal Protein Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Dodavatel: Bioss
BS-7866R-A680
BOSSBS-7866R-A680EA 25430 CZK
BOSSBS-7866R-A680
Anti-Intra Acrosomal Protein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Protilátky
This gene encodes a testis-specific, differentiation antigen, acrosomal vesicle protein 1, that arises within the acrosomal vesicle during spermatogenesis, and is associated with the acrosomal membranes and matrix of mature sperm. The acrosomal vesicle protein 1 may be involved in sperm-zona binding or penetration. Alternatively spliced transcript variants have been described.

Type: Primary
Antigen: Intra Acrosomal Protein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 171-265/265
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR