Tisk…

Anti-SPAG4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Dodavatel: Bioss
BS-7866R-A555
BOSSBS-7866R-A555EA 25430 CZK
BOSSBS-7866R-A555
Anti-SPAG4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
This gene encodes a testis-specific, differentiation antigen, acrosomal vesicle protein 1, that arises within the acrosomal vesicle during spermatogenesis, and is associated with the acrosomal membranes and matrix of mature sperm. The acrosomal vesicle protein 1 may be involved in sperm-zona binding or penetration. Alternatively spliced transcript variants have been described. [provided by RefSeq, Nov 2010].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPAG4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu SPAG4
Environmentally Preferable
Název antigenu Sperm Associated Antigen 4
Synonyma antigenu SUN4|CT127
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 555
Reaktivita Human
Hostitel Rabbit
Identifikační číslo genu 6676
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at –20 °C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR