Tisk…
Anti-DUSP13 Rabbit Polyclonal Antibody (Cy7®)
Dodavatel: Bioss
BS-7797R-CY7
BOSSBS-7797R-CY7EA 25630 CZK
BOSSBS-7797R-CY7
Anti-DUSP13 Rabbit Polyclonal Antibody (Cy7®)
Protilátky
Members of the protein-tyrosine phosphatase superfamilycooperate with protein kinases to regulate cell proliferation anddifferentiation. This superfamily is separated into two familiesbased on the substrate that is dephosphorylated. One family, thedual specificity phosphatases (DSPs) acts on both phosphotyrosineand phosphoserine/threonine residues. This gene encodes differentbut related DSP proteins through the use of non-overlapping openreading frames, alternate splicing, and presumed differenttranscription promoters. Expression of the distinct proteins fromthis gene has been found to beMore Product Information