Tisk…

Anti-DSN1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Dodavatel: Bioss
BS-7796R-A350
BOSSBS-7796R-A350EA 38440 CZK
BOSSBS-7796R-A350
Anti-DSN1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
This gene encodes a kinetochore protein that functions aspart of the minichromosome instability-12 centromere complex. Theencoded protein is required for proper kinetochore assembly andprogression through the cell cycle. Alternative splicing results inmultiple transcript variants. [provided by RefSeq, Feb 2009].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DSN1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR