Tisk…

Anti-KIF20A Rabbit Polyclonal Antibody (Cy7®)

Dodavatel: Bioss
BS-7750R-CY7
BOSSBS-7750R-CY7EA 25630 CZK
BOSSBS-7750R-CY7
Anti-KIF20A Rabbit Polyclonal Antibody (Cy7®)
Protilátky
Mitotic kinesin required for chromosome passenger complex (CPC)-mediated cytokinesis. Following phosphorylation by PLK1, involved in recruitment of PLK1 to the central spindle. Interacts with guanosine triphosphate (GTP)-bound forms of RAB6A and RAB6B. May act as a motor required for the retrograde RAB6 regulated transport of Golgi membranes and associated vesicles along microtubules. Has a microtubule plus end-directed motility.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KIF20A
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
More Product Information
Objednat

Produktová specifikace

Symbol antigenu KIF20A
Název antigenu Kinesin Family Member 20A
Environmentally Preferable
Synonyma antigenu MKLP2|GG10_2|mitotic kinesin-like protein 2|rabkinesin-6|RAB6KIFL|rab6-interacting kinesin-like protein|kinesin-like protein KIF20A|kinesin-like (rabkinesin6)|RAB6 interacting
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy7®
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 10112
Izotyp IgG
Imunochemie Yes
Imunogen 370-420/890
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at –20 °C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR