Tisk…
Anti-BCAM Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Dodavatel: Bioss
BS-7546R-A350
BOSSBS-7546R-A350EA 25430 CZK
BOSSBS-7546R-A350
Anti-BCAM Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
This gene encodes Lutheran blood group glycoprotein, a member of the immunoglobulin superfamily and a receptor for the extracellular matrix protein, laminin. The protein contains five extracellular immunoglobulin domains, a single transmembrane domain, and a short C-terminal cytoplasmic tail. This protein may play a role in epithelial cell cancer and in vaso-occlusion of red blood cells in sickle cell disease. Polymorphisms in this gene define some of the antigens in the Lutheran system and also the Auberger system. Inactivating variants of this gene result in the recessive Lutheran nullMore Product Information