Tisk…

Anti-S1PR3 Rabbit Polyclonal Antibody (Cy5.5®)

Dodavatel: Bioss
BS-7541R-CY5.5
BOSSBS-7541R-CY5.5EA 25630 CZK
BOSSBS-7541R-CY5.5
Anti-S1PR3 Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
Receptor for the lysosphingolipid sphingosine 1-phosphate (S1P). S1P is a bioactive lysophospholipid that elicits diverse physiological effect on most types of cells and tissues. When expressed in rat HTC4 hepatoma cells, is capable of mediating S1P-induced cell proliferation and suppression of apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: S1PR3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Produktová specifikace

Symbol antigenu S1PR3
Environmentally Preferable
Název antigenu Sphingosine-1-phosphate Receptor 3
Synonyma antigenu S1P3|EDG-3|EDG3|LPB3
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy5.5®
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 1903
Izotyp IgG
Imunochemie Yes
Imunogen 175-205/378
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at –20 °C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR