Tisk…
Anti-TROVE2 Rabbit Polyclonal Antibody
Dodavatel: Bioss
BS-7116R
BOSSBS-7116REA 20650 CZK
BOSSBS-7116R
Anti-TROVE2 Rabbit Polyclonal Antibody
Protilátky
Ro autoantigens are of clinical significance because directed against them are found in most patients with primary Sjqgren syndrome, subacute cutaneous lupus erythematosus (SLE), neonatal lupus erythematosus, ANA-negative lupus erythematosus, and systemic lupus erythematosus-like disease secondary to homozygous C2 or C4 complement deficiency (1). Ro/SSA is a ribonucleoprotein that binds to auto in 35 to 50% of patients with SLE and in up to 97% of patients with Sjqgren syndrome (2). The Ro/SSA particle consists of a single immunoreactive protein noncovalently bound with one of four small RNAMore Product Information