Tisk…

Anti-OCIAD1 Rabbit Polyclonal Antibody

Dodavatel: Bioss
BS-7001R
BOSSBS-7001REA 20650 CZK
BOSSBS-7001R
Anti-OCIAD1 Rabbit Polyclonal Antibody
Protilátky
Tissue specificity; Isoform 1 is highly expressed in many tissues, including testis, brain, placenta, ovary, prostate and mammary gland. Isoform 2 expression is restricted to the central nervous system including brain, cerebellum and spinal cord.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: OCIAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR